Tisk…

Anti-PXN Mouse Monoclonal Antibody [clone: PXC-10]

ABCAAB11101-100EA 16280 CZK
ABCAAB11101-100
Anti-PXN Mouse Monoclonal Antibody [clone: PXC-10]
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone: PXC-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR