Tisk…

Anti-Synaptobrevin Mouse Monoclonal Antibody [clone: SP11]

ABCAAB11104-100EA 17330 CZK
ABCAAB11104-100
Anti-Synaptobrevin Mouse Monoclonal Antibody [clone: SP11]
Type: Primary
Antigen: Synaptobrevin
Clonality: monoclonal
Clone: SP11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR