Tisk…

Anti-APP Rabbit Polyclonal Antibody

ABCAAB11119-50EA 15650 CZK
ABCAAB11119-50
Anti-APP Rabbit Polyclonal Antibody
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR