Tisk…

Anti-PSEN2 Goat Polyclonal Antibody

ABCAAB11148-50EA 15650 CZK
ABCAAB11148-50
Anti-PSEN2 Goat Polyclonal Antibody
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR