Tisk…

Anti-THY1 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: F15-42-1]

ABCAAB11155-500EA 7470 CZK
ABCAAB11155-500
Anti-THY1 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: F15-42-1]
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: F15-42-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR