Tisk…

Anti-MBP Mouse Monoclonal Antibody [clone: 2]

ABCAAB11159-1EA 17250 CZK
ABCAAB11159-1
Anti-MBP Mouse Monoclonal Antibody [clone: 2]
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: monoclonal
Clone: Clone 2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR