Tisk…

Anti-TRPM2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB11168-50EA 27640 CZK
ABCAAB11168-50
Anti-TRPM2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRPM2 (transient receptor potential cation channel, subfamily M, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR