Tisk…

Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11+PCK-26+CY-90+ KS-1A3+ M20+A53-B/A2]

ABCAAB11213-200EA 16280 CZK
ABCAAB11213-200
Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11+PCK-26+CY-90+ KS-1A3+ M20+A53-B/A2]
Type: Primary
Antigen: pan Cytokeratin
Clonality: monoclonal
Clone: C-11+PCK-26+CY-90+ KS-1A3+ M20+A53-B/A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR