Tisk…

Anti-beta I + II Tubulin Mouse Monoclonal Antibody [clone: JDR.3B8]

ABCAAB11313-100EA 16080 CZK
ABCAAB11313-100
Anti-beta I + II Tubulin Mouse Monoclonal Antibody [clone: JDR.3B8]
Type: Primary
Antigen: beta I + II Tubulin
Clonality: monoclonal
Clone: JDR.3B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR