Tisk…

Anti-GHR Mouse Monoclonal Antibody [clone: MAB 263]

ABCAAB11380-100EA 16480 CZK
ABCAAB11380-100
Anti-GHR Mouse Monoclonal Antibody [clone: MAB 263]
Type: Primary
Antigen: GHR (growth hormone receptor)
Clonality: monoclonal
Clone: MAB 263
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR