Tisk…

Anti-hCG beta core fragment Mouse Monoclonal Antibody [clone: INN-hCG-106]

Dodavatel: Abcam
ABCAAB11382-250EA 21300 CZK
ABCAAB11382-250
Anti-hCG beta core fragment Mouse Monoclonal Antibody [clone: INN-hCG-106]
Protilátky
Type: Primary
Antigen: hCG beta core fragment
Clonality: monoclonal
Clone: INN-hCG-106
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR