Tisk…

Anti-CD3 zeta Mouse Monoclonal Antibody FITC (Fluorescein) [clone: G3]

ABCAAB11505-500EA 6870 CZK
ABCAAB11505-500
Anti-CD3 zeta Mouse Monoclonal Antibody FITC (Fluorescein) [clone: G3]
Type: Primary
Antigen: CD3 zeta
Clonality: monoclonal
Clone: G3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR