Tisk…

Anti-ADAM12 Rabbit Polyclonal Antibody

ABCAAB11536-25EA 15650 CZK
ABCAAB11536-25
Anti-ADAM12 Rabbit Polyclonal Antibody
Type: Primary
Antigen: ADAM12 (ADAM metallopeptidase domain 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR