Tisk…

Anti-BSG Goat Polyclonal Antibody

ABCAAB11572-25EA 16760 CZK
ABCAAB11572-25
Anti-BSG Goat Polyclonal Antibody
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR