Tisk…

Anti-PCOLE2 Rabbit Polyclonal Antibody

ABCAAB11578-25EA 15650 CZK
ABCAAB11578-25
Anti-PCOLE2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PCOLE2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR