Tisk…

Anti-IgG1 Rat Monoclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB11587-250EA 12660 CZK
ABCAAB11587-250
Anti-IgG1 Rat Monoclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR