Tisk…

Anti-IgG2b Rat Monoclonal Antibody [clone: LO-MG2b-2] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abcam
ABCAAB11655-250EA 12140 CZK
ABCAAB11655-250
Anti-IgG2b Rat Monoclonal Antibody [clone: LO-MG2b-2] (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: monoclonal
Clone: LO-MG2b-2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR