Tisk…

Anti-IgG2c Mouse Monoclonal Antibody [clone: MARG2c-5] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abcam
ABCAAB11677-250EA 12140 CZK
ABCAAB11677-250
Anti-IgG2c Mouse Monoclonal Antibody [clone: MARG2c-5] (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: Rat IgG2c
Clonality: monoclonal
Clone: MARG2c-5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR