Tisk…

Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10.7]

ABCAAB11841-6EA 16080 CZK
ABCAAB11841-6
Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10.7]
Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: monoclonal
Clone: 4B10.7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR