Tisk…

Anti-Interferon gamma Mouse Monoclonal Antibody [clone: F14]

ABCAAB11866-50EA 15370 CZK
ABCAAB11866-50
Anti-Interferon gamma Mouse Monoclonal Antibody [clone: F14]
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: F14
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR