Tisk…

Anti-C3 / C3a Mouse Monoclonal Antibody [clone: 474]

Dodavatel: Abcam
ABCAAB11872-50EA 22490 CZK
ABCAAB11872-50
Anti-C3 / C3a Mouse Monoclonal Antibody [clone: 474]
Protilátky
Type: Primary
Antigen: C3 / C3a
Clonality: monoclonal
Clone: 474
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR