Tisk…

Anti-C5 / C5a Mouse Monoclonal Antibody [clone: 2942]

Dodavatel: Abcam
ABCAAB11877-50EA 22860 CZK
ABCAAB11877-50
Anti-C5 / C5a Mouse Monoclonal Antibody [clone: 2942]
Protilátky
Type: Primary
Antigen: C5 / C5a
Clonality: monoclonal
Clone: 2942
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR