Tisk…

Anti-Menin Rabbit Polyclonal Antibody

ABCAAB11893-50EA 16280 CZK
ABCAAB11893-50
Anti-Menin Rabbit Polyclonal Antibody
Type: Primary
Antigen: Menin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR