Tisk…

Anti-ATF6 Mouse Monoclonal Antibody [clone: 70B1413]

ABCAAB11909-100EA 18560 CZK
ABCAAB11909-100
Anti-ATF6 Mouse Monoclonal Antibody [clone: 70B1413]
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: monoclonal
Clone: 70B1413
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR