Tisk…

Anti-KDR Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB11939-100EA 23720 CZK
ABCAAB11939-100
Anti-KDR Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR