Tisk…

Anti-FOSB Mouse Monoclonal Antibody [clone: 83B1138]

Dodavatel: Abcam
ABCAAB11959-50EA 23780 CZK
ABCAAB11959-50
Anti-FOSB Mouse Monoclonal Antibody [clone: 83B1138]
Protilátky
Type: Primary
Antigen: FOSB (FBJ murine osteosarcoma viral oncogene homolog B)
Clonality: monoclonal
Clone: 83B1138
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR