Tisk…

Anti-RNA polymerase beta Mouse Monoclonal Antibody [clone: 8RB13]

ABCAAB12087-50EA 16880 CZK
ABCAAB12087-50
Anti-RNA polymerase beta Mouse Monoclonal Antibody [clone: 8RB13]
Type: Primary
Antigen: RNA polymerase beta
Clonality: monoclonal
Clone: 8RB13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR