Tisk…

Anti-PIN1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB12107-100EA 0 CZK
ABCAAB12107-100
Anti-PIN1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR