Tisk…

Anti-STAT5A Rabbit Polyclonal Antibody

ABCAAB12113-50EA 15650 CZK
ABCAAB12113-50
Anti-STAT5A Rabbit Polyclonal Antibody
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR