Tisk…

Anti-RELA Rabbit Polyclonal Antibody

ABCAAB12146-50EA 15650 CZK
ABCAAB12146-50
Anti-RELA Rabbit Polyclonal Antibody
Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR