Tisk…

Anti-PTPN5 Rabbit Polyclonal Antibody

ABCAAB12151-100EA 15650 CZK
ABCAAB12151-100
Anti-PTPN5 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PTPN5 (protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR