Tisk…

Anti-HDAC4 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB12172-50EA 28410 CZK
ABCAAB12172-50
Anti-HDAC4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HDAC4 (histone deacetylase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR