Tisk…

Anti-Metallothionein Mouse Monoclonal Antibody [clone: UC1MT]

Dodavatel: Abcam
ABCAAB12228-100EA 23960 CZK
ABCAAB12228-100
Anti-Metallothionein Mouse Monoclonal Antibody [clone: UC1MT]
Protilátky
Type: Primary
Antigen: Metallothionein
Clonality: monoclonal
Clone: UC1MT
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR