Tisk…
Anti-PSEN1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB12272-100EA 26350 CZK
ABCAAB12272-100
Anti-PSEN1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat