Tisk…

Anti-RBM14 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB12325-50EA 27760 CZK
ABCAAB12325-50
Anti-RBM14 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RBM14 (RNA binding motif protein 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR