Tisk…

Anti-FHL2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB12328-50EA 23040 CZK
ABCAAB12328-50
Anti-FHL2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FHL2 (four and a half LIM domains 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR