Tisk…

Anti-Macrophage Inflammatory Protein 4 Rabbit polyclonal antibody Biotin

ABCAAB12749-25EA 15370 CZK
ABCAAB12749-25
Anti-Macrophage Inflammatory Protein 4 Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: Macrophage Inflammatory Protein 4
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR