Tisk…

Anti-SYK Rabbit polyclonal antibody

ABCAAB12862-50EA 16280 CZK
ABCAAB12862-50
Anti-SYK Rabbit polyclonal antibody
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR