Tisk…

Anti-HIST2H2BE Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB12929-50EA 23870 CZK
ABCAAB12929-50
Anti-HIST2H2BE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HIST2H2BE (histone cluster 2, H2be)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR