Tisk…

Anti-Synaptotagmin Mouse Monoclonal Antibody [clone: ASV30]

Dodavatel: Abcam
ABCAAB13259-100EA 23530 CZK
ABCAAB13259-100
Anti-Synaptotagmin Mouse Monoclonal Antibody [clone: ASV30]
Protilátky
Type: Primary
Antigen: Synaptotagmin
Clonality: monoclonal
Clone: ASV30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR