Tisk…

Anti-RIF1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB13422-100EA 22860 CZK
ABCAAB13422-100
Anti-RIF1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RIF1 (RAP1 interacting factor homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR