Tisk…

Anti-HSP90B1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB13509-100EA 23570 CZK
ABCAAB13509-100
Anti-HSP90B1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HSP90B1 (heat shock protein 90kDa beta (Grp94), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR