Tisk…

Anti-AIRE Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB13573-100EA 22130 CZK
ABCAAB13573-100
Anti-AIRE Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR