Tisk…

Anti-DAP Kinase 2 Rabbit polyclonal antibody

ABCAAB13656-100EA 15370 CZK
ABCAAB13656-100
Anti-DAP Kinase 2 Rabbit polyclonal antibody
Type: Primary
Antigen: DAP Kinase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR