Tisk…

Anti-PNMA2 Rabbit polyclonal antibody

ABCAAB13705-200EA 15370 CZK
ABCAAB13705-200
Anti-PNMA2 Rabbit polyclonal antibody
Type: Primary
Antigen: PNMA2 (paraneoplastic Ma antigen 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR