Tisk…

Anti-ASB11 Rabbit polyclonal antibody

ABCAAB13711-100EA 15370 CZK
ABCAAB13711-100
Anti-ASB11 Rabbit polyclonal antibody
Type: Primary
Antigen: ASB11 (ankyrin repeat and SOCS box containing 11, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR