Tisk…

Anti-SMAD5 Rabbit polyclonal antibody

ABCAAB13724-200EA 18390 CZK
ABCAAB13724-200
Anti-SMAD5 Rabbit polyclonal antibody
Type: Primary
Antigen: SMAD5 (SMAD family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR