Tisk…

Anti-RGS11 Rabbit polyclonal antibody

ABCAAB13725-200EA 16360 CZK
ABCAAB13725-200
Anti-RGS11 Rabbit polyclonal antibody
Type: Primary
Antigen: RGS11 (regulator of G-protein signaling 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR