Tisk…

Anti-SETD7 Rabbit polyclonal antibody

ABCAAB13731-200EA 17330 CZK
ABCAAB13731-200
Anti-SETD7 Rabbit polyclonal antibody
Type: Primary
Antigen: SETD7 (SET domain containing (lysine methyltransferase) 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR