Tisk…

Anti-IL2 Mouse monoclonal antibody [clone: N7.48A]

ABCAAB13777-100EA 15650 CZK
ABCAAB13777-100
Anti-IL2 Mouse monoclonal antibody [clone: N7.48A]
Type: Primary
Antigen: IL2 (interleukin 2)
Clonality: monoclonal
Clone: N7.48A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR